Learn More
Bel-Art™ SP Scienceware™ Lab-Aire™ II Glassware Dryers Accessories
Replacement pegs for use with Lab-Aire II benchtop and wallmount dryer models.
34.35 € - 36.95 €
Specifications
Product Type Replacement Peg
Autoclavable Not Autoclavable
For Use With (Equipment) Lab-Aire II benchtop and wallmount dryers
View More Specs
Products 3
Product Code Brand Length (English) Size Price Quantity & Availability  
Product Code Brand Length (English) Size Price Quantity & Availability  
13293409
View Documents
Bel-Art™
F18931-0002
4 in. Medium
36.85 €
Pack of 19
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
13213419
View Documents
Bel-Art™
F18931-0001
3 in. Small
34.35 €
Pack of 19
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
13283409
View Documents
Bel-Art™
F18931-0003
5 in. Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
    • Available as small (3 in.), medium (4 in.) and large (5 in.) pegs
    • Pegs can be moved and repositioned to hold variety of labware including beakers, flasks, bottles, test tubes and cylinders
Specifications
Specifications
Replacement Peg
Not Autoclavable
Lab-Aire II benchtop and wallmount dryers