Learn More
Bel Art™ Acid/Solvent Bottle Carriers
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles
Brand: Bel Art™ F16959-0000
153.55 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- Offer shatter resistant protection for glass bottles
- Cone style lid has a 0.625 in. (16 mm) opening in top for insertion of tubing; simplifies the transfer of liquids without pouring
- Truncated style lid permits pouring without the removal of the lid
- Designed to hold standard US reagent bottles
Specifications
Bottle Carrier | |
Not Autoclavable | |
Bottles | |
Conical | |
No | |
To hold standard US reagent bottles | |
32 cm | |
32 cm | |
160 mm | |
18cm |
Yes | |
White | |
Polyethylene | |
None | |
Non-sterile | |
1 | |
18 cm | |
12.6 in. | |
180 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.