missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Acid/Solvent Bottle Carriers
Polyethylene carriers with epoxy coated, heavy-gauge steel wire bails have wide ribs in both the sides and on the bottom to support and cushion bottles
116.00 € - 175.00 €
Specifications
Product Type | Bottle Carrier |
---|---|
Autoclavable | Not Autoclavable |
Shelves | None |
Stackable | No |
Sterility | Non-sterile |
Product Code | Brand | Height (English) | Height (Metric) | Material | Holds | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Height (Metric) | Material | Holds | Price | Quantity & Availability | |||
11733207
|
Bel Art™
F16958-0000 |
14.37 in. | 37 cm | Polyethylene | Bottles (4 L) |
125.91 €
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||
11753207
|
Bel Art™
F16960-0000 |
14.37 in. | 37 cm | Polypropylene | Bottles (4 L) |
126.00 €
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
|||
11723207
|
Bel Art™
F16957-0000 |
13.5 in. | 35 cm | Polypropylene | Bottles (2.5 L) |
116.00 €
Each |
Estimated Shipment: 05-06-2025
Log in to see stock. |
|||
11743207
|
Bel Art™
F16959-0000 |
12.6 in. | 32 cm | Polyethylene | Bottles |
175.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||
Description
- Offer shatter resistant protection for glass bottles
- Cone style lid has a 0.625 in. (16 mm) opening in top for insertion of tubing; simplifies the transfer of liquids without pouring
- Truncated style lid permits pouring without the removal of the lid
- Designed to hold standard US reagent bottles
Specifications
Bottle Carrier | |
None | |
Non-sterile |
Not Autoclavable | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel Art™ Acid/Solvent Bottle Carriers