Learn More
Bel-Art™ Air Operated Turbine Magnetic Stirrer
Compact, small footprint stirrer is optimal for stirring vessels up to 1L
Brand: Bel-Art™ F37002-0000
75.19 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Includes: Tubing is supplied to connect to an air supply.
Descripción
- Uses only 3 to 8 psi (21 to 55 kPa) to drive the magnetic stirrer
- Excellent for use where potentially explosive solvent vapor are present
- Can be used with magnetic stirring bars up to 2.5 cm (1 in.) in length
- Eliminates sparking hazard from electrical sources
- Made from high impact polystyrene to ensure long life and easy clean-up in case of spills
Especificaciones
0 to 2000 rpm | |
Polystyrene | |
None | |
0/1 V | |
4 in. | |
0.75 in. | |
Air Operated Turbine Magnetic Stirrer |
1 | |
1 L | |
Round | |
4 x 0.75 in. (102 x 19 mm) | |
102 mm | |
19 mm |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.