Learn More
Bel-Art™ Cloning Cylinders
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
Brand:  Bel-Art™ F37847-0300
 View more versions of this product
Product Code. 10246182
  • 114.27 € / Pack of 50
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
  • Tapered
  • Good sealing surface
  • Polystyrene
  • Sterile
Order Info
Individual sizes supplied five bags of 10 cylinders each; Variety Pack contains four bags with 10 of each size
Specifications
Specifications
Polystyrene
11 mm
0.37 in.
0.443 in.
Large
9.5 mm
Sterile
Cloning Cylinder
Individual clone isolation and propagation