Learn More
Bel-Art™ Cloning Cylinders
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
Brand:  Bel-Art™ H37847-0000
 View more versions of this product
Product Code. 11795176
  • 151.00 € / Pack of 40
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from polystyrene. SP Scienceware™ Cloning Cylinders facilitates the isolation of individual cell clones.
  • Tapered
  • Good sealing surface
  • Polystyrene
  • Sterile
Order Info
Individual sizes supplied five bags of 10 cylinders each; Variety Pack contains four bags with 10 of each size
Specifications
Specifications
Polystyrene
Sterile
Cloning Cylinder
Small, Medium, Large (Variety)
4.7, 6.4, 9.5 mm
0.185 in., 0.251 in., 0.37 in.
Isolation of individual cell clones