Learn More
Bel-Art™ SP Scienceware™ Magnetic Bead Separation Racks
Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays.
Brand:  Bel-Art™ F19900-0002
 View more versions of this product
Product Code. 16650584
  • 462.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Alert:
These separation racks contain powerful Neodymium magnets. Misuse can result in injury.
  • Do not use this product if you have a pacemaker or any other medical magnetized implant
  • Keep away from tools and objects that could be damaged by the magnetic field
  • Racks should be handled and stored away from devices subject to magnetic damage including credit cards and other products containing magnetic recording devices
  • Keep away from delicate instruments, watches, electronic equipment, displays and monitors
  • Magnets may attract steel or other magnetic material with high mechanical forces
  • Avoid contact between two magnets
  • Do not pull the magnets apart if contact has been made; twist off to prevent damage to the unit or fingers
Description
Description
  • Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves of 1.5 to 50ml models allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
  • 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes 
  • Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96 etc.
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents
Specifications
Specifications
Magnetic Bead Separation Rack
Microcentrifuge Tubes (1.5 to 2 mL)
10
Clear
Neodymium Magnets, Acrylic
General Laboratories, Immunology, Proteomics, Genomics, Separation Methods