missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Magnetic Bead Separation Racks

Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays.

462.00 € - 483.76 €

Specifications

Product Type Magnetic Bead Separation Rack
Color Clear
Material Neodymium Magnets, Acrylic
For Use With (Application) General Laboratories, Immunology, Proteomics, Genomics, Separation Methods
View More Specs

Alert:

These separation racks contain powerful Neodymium magnets. Misuse can result in injury.

  • Do not use this product if you have a pacemaker or any other medical magnetized implant
  • Keep away from tools and objects that could be damaged by the magnetic field
  • Racks should be handled and stored away from devices subject to magnetic damage including credit cards and other products containing magnetic recording devices
  • Keep away from delicate instruments, watches, electronic equipment, displays and monitors
  • Magnets may attract steel or other magnetic material with high mechanical forces
  • Avoid contact between two magnets
  • Do not pull the magnets apart if contact has been made; twist off to prevent damage to the unit or fingers


Products 3
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
16640584
View Documents
Bel-Art™
F19900-0001
Tubes (5 or 15 mL)
462.00 €
Each
Estimated Shipment: 26-02-2025
to see stock.
Add to basket
 
17840839
View Documents
Bel-Art™
F19900-0003
PCR Tube Plate (96-well)
483.76 €
Each
Please to purchase this item. Need a web account? Register with us today!
16650584
View Documents
Bel-Art™
F19900-0002
Microcentrifuge Tubes (1.5 to 2 mL)
462.00 €
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
Description

Description

  • Powerful racks sequester magnetic beads so that affinity ligands or rinse solutions can be added or decanted in assays
  • Powerful magnets quickly secure beads
  • Spring plunger on tube sleeves of 1.5 to 50ml models allows beads to be sequestered at various heights along the tube wall
  • Simply turn tube for efficient bead mixing
  • 96-well PCR plate model has 24 magnets that snugly hold the plate while attracting beads to the wall of the tubes 
  • Flat bottom microplate model has magnets covering entire footprint to accept all standard micro-plates with any number of wells; 6, 12, 24, 48, 96 etc.
  • Racks fit microplates following ANSI standards
  • Clear acrylic allows unobstructed view of tube contents
Specifications

Specifications

Magnetic Bead Separation Rack
Clear
Neodymium Magnets, Acrylic
General Laboratories, Immunology, Proteomics, Genomics, Separation Methods
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.