Learn More
Bel-Art™ Mini Sampler Spoons
Avoid cross contamination by using a different spoon for each sample
Brand: Bel-Art™ 06263-50
This item is not returnable.
View return policy
Description
- These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
- Made from white high-impact polystyrene plastic
- All sterile products are lot stamped for accurate tracing
Specifications
Spoon | |
Not Autoclavable | |
Single-use | |
White |
Polystyrene | |
0.05 mL | |
Sterile | |
Designed for retrieving small, specific volume samples of powders or similar materials |