Learn More
Bel-Art™ SP Scienceware™ Mini Sampler Spoons
Avoid cross contamination by using a different spoon for each sample
Brand:  Bel-Art™ F36721-0025
 View more versions of this product
Product Code. 15580102
  • 25.71 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • These disposable mini measures are designed for retrieving small, specific volume samples of powders or similar materials
  • Made from white high-impact polystyrene plastic
  • All sterile products are lot stamped for accurate tracing
Specifications
Specifications
Spoon
0.25 mL
Sterile
Designed for retrieving small, specific volume samples of powders or similar materials
Polystyrene
Single-use
White