missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid™ Half-Size Test-Tube Racks

For use in water baths, laminar flow hoods, fume hoods or any place where space is limited

Brand:  Bel-Art™ F18788-1600

 View more versions of this product

Product Code. 15590042

  • 47.85 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Epoxy coating for chemical and corrosion resistance
  • Square holes in the grid
  • Three decks with slotted grid bottom
  • Available to hold 10-13, 15-16 or 18-20mm diameter tubes
  • Autoclavable
  • Unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand temperature range of −223°C (−369°F) to 121°C (250°F)
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (13 to 16 mm)
24
13 - 16Tubes
4.9 x 3.9 x 2.5 in. (12.5 x 9.9 x 6.4cm)
6 x 4
Green
Epoxy-coated Steel
Non-sterile
-223°C to +121°C
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.