Learn More
Bel-Art™ SP Scienceware™ Secador™ Desiccator Replacement Shelves
Convert 4.0 units from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
Brand:  Bel-Art™ F42074-1001
 View more versions of this product
Product Code. 11832732
  • 68.31 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fit auto and nonelectric models
  • Durable, clear polystyrene construction
  • Perforated to faciilitate air flow
Specifications
Specifications
Desiccator Shelf (Vertical)
Polystyrene
Convert 4.0 units from horizontal to vertical desiccator type