Learn More
Bel-Art™ SP Scienceware™ Secador™ Desiccator Replacement Shelves
Convert 4.0 units from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
49.80 € - 68.31 €
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
12388995
|
Bel-Art™
F42074-0001 |
Desiccator Shelf (Horizontal) | Convert 4.0 units from vertical to horizontall desiccator type |
49.80 €
Each |
Estimated Shipment: 12-03-2025
Log in to see stock. |
||||
11832732
|
Bel-Art™
F42074-1001 |
Desiccator Shelf (Vertical) | Convert 4.0 units from horizontal to vertical desiccator type |
68.31 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
||||
Description
- Fit auto and nonelectric models
- Durable, clear polystyrene construction
- Perforated to faciilitate air flow
Specifications
Polystyrene |