Learn More
Bel-Art™ Secador™ Mini Desiccator Cabinet with Gas Ports
The sturdy construction and compact size of this desiccator cabinet assures reliable storage for moisture sensitive products when benchtop space is at a premium.
Brand:  Bel-Art™ F42075-1002
Product Code. 11872742
  • 361.00 € / Each
Estimated Shipment: 31-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Large side swinging door maximizes access to interior space.
  • Features patented seal design with tab latches and security loop for padlock or tamper-evident seals.
  • Opens right to left.
  • Top mounted gas ports with barbed fittings accept 0.25 in. I.D. tubing.
  • Easy-to-view dial hygrometer located in the door front.
  • Clear polystyrene for easy viewing of contents.
  • Gas ports can be closed to use desiccator with standard desiccant cartridges.
  • 10 D x 13.3 W x 14 in. H (25.4 x 33.8 x 35.6 cm).
  • Mini desiccator model is molded of polystyrene and does not block UV light.
  • For UV protection, select a Secador 1.0 model or larger.
Specifications
Specifications
Clear
Copolyester, Polystyrene
Latched, swinging
Fitting, stopcock (for 0.25 in. I.D. tubing) and instructions, top mounted two gas ports, locking loop with lock or tamper-evident seal, door-mounted dial hygrometer
13.3 in.
33.8 cm
8.5 in.
21.6 cm
25.4 cm
Front-loading
Desiccator
Assures reliable storage for moisture sensitive products when benchtop space is at a premium
0.35 cu. ft.
9.3 in.
23.6 cm
5 in.
10 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ Secador™ Mini Desiccator Cabinet with Gas Ports > Mini Desiccator Cabinet; w/Gas Ports