missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Acrylic Desiccator Cabinets
GREENER_CHOICE

Store anhydrous, hygroscopic biological and chemical compounds in a tight seal with clear Thermo Scientific™ Nalgene™ Acrylic Desiccator Cabinets.

Brand:  Thermo Scientific™ 5317-0120

762.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10450723

  • 793.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Store any components requiring controlled storage conditions, such as anhydrous and hygroscopic biological and chemical compounds and electronic assemblies. Thermo Scientific™ Nalgene™ Acrylic Desiccator Cabinets are clear and feature gaskets to provide a tight seal.

Features:

  • Neoprene gasket on door ensures tight seal
  • Door is secured with two (three on largest size) stainless-steel latches and full-height hinge
  • Removable, lipped tray holds solid desiccant in place
  • Shelves are removable, adjustable and vented to maximize airflow

Ordering Information:
All cabinets measure 12L x 12 in.W (30.5 x 30.5cm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Front Loading
5
30.5 cm
Desiccator Cabinet Acrylic 305 x 305 x 305mm
3
12 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Schematics & Diagrams

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Acrylic Desiccator Cabinets > Desiccator Cabinets; Height: 12 in. (30.5cm); No. of Shelves: 3

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.