missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Single Scale Graduated Cylinders

With raised numerals for quick, accurate reading

Brand:  Bel-Art™ F28454-0000

 View more versions of this product

Product Code. 30193444

  • 10.46 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flared top rims and large spouts facilitate pouring
  • Octagonal shaped bases prevent tipping and rolling
  • Non-wetting polypropylene plastic surface eliminates concave meniscus; makes reading easier and more accurate
  • Autoclavable at 121°C (250°F)

Specifications

Specifications

50 mL
1 in.
7.9 in.
Autoclavable
Round
7.9 in.
Graduated Cylinder
Polypropylene
Single
1 mL
1mL
Octagonal
20.1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.