missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Tech Pens

Heavy-duty soft tip disposable pens come in seven colors and write on glass, metal, ceramics, plastics, paper and more

Brand:  Bel-Art™ F13384-0006

 View more versions of this product

Product Code. 10133392

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Oil-based marking paint resists washing, weathering, mild acids and alkalis
  • Operate within the temperature range of -46™ to +65°C (-50° to +150°F)
  • Withstands autoclaving 121°C (250°F)
  • Mark is approximately 1mm (0.040 in.) wide

Specifications

Specifications

Soft Tip
Green
Single-use
Writes on Glass, Metal, Ceramics, Plastics, Paper etc
-46°C to +65°C
1 mm
Technical Pen
Autoclavable
Snap cap
-50°F to +150°F
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.