Learn More
Bel-Art™ SP Scienceware™ Varipet™ Pipettor
Self-filling pipetting aid designed for speed, ease, and convenience in a dependable, positive displacement dispenser
Brand:  Bel-Art™ F37895-1100
 View more versions of this product
Product Code. 10374415
  • 372.00 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene handle to permit extended use without fatigue
  • PTFE plunger assures smooth, consistent operation in repetitive tasks
  • Dispensing volume can be adjusted by turning the knob at the base of the unit
  • Accuracy is ±0.5%
Teflon™ Tip
With chemical resistance because the fluids only contact the borosilicate glass and Teflon
Luer Lock
Supplied with chrome-plated steel Luer lock fitting, complete with removable cannula for general use
Specifications
Specifications
±0.5%
1
10 mL
0.2mL
Luer lock tip
Pipette
Polypropylene handle, PTFE plunger, Borosilicate Glass syringe, Chrome plated Steel luer lock fitting