missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Bench-Top Mini-Bottle Roller Apparatus
For cell cultivation research or staining procedures. Wheaton™ Bench-Top Mini-Bottle Roller Apparatus allows for serum and media bottles, as well as culture tubes.
2770.00 €
Specifications
Speed Range | 3 to 45 rpm |
---|---|
Electrical Requirements | 100/240 V, 50/60 Hz, 14 W |
Voltage | 100/240 V |
Temperature Range (Metric) | 40°C |
Wattage | 14 W |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
12412082
|
Wheaton™
W348923-D |
D Model |
2770.00 €
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
Description
- Accommodates bottles 38 to 60mm in diameter and 240mm long
- Deck kits available to double or triple capacity
- Precise and reproducible speed adjustments
Specifications
3 to 45 rpm | |
100/240 V | |
14 W | |
50/60 Hz | |
18 cm | |
8 kg | |
1 Ea. |
100/240 V, 50/60 Hz, 14 W | |
40°C | |
CE, CSA, UL | |
32.5 cm | |
32 cm | |
For the growth and observation of various cell cultures |