Learn More
Thermo Scientific™ Benchtop Coolers
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
218.00 € - 465.00 €
Product Code | Brand | Temperature Range | Array | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range | Array | Holds | Price | Quantity & Availability | ||||
10777232
![]() |
Thermo Scientific™
355501 |
-20°C | 4 x 5 | 20 Tubes (0.2 to 0.5 mL) |
319.00 €
Each |
In Stock
Log in to see stock. |
||||
10292581
![]() |
Thermo Scientific™
5115-0012 |
-20°C | 3 x 4 | 12 Tubes (0.2 to 0.5 mL) |
218.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||
10252631
![]() |
Thermo Scientific™
5115-0032 |
-20°C | 4 x 8 | 32 Tubes (0.2 to 0.5 mL) |
418.00 €
Each |
In Stock
Log in to see stock. |
||||
10525021
![]() |
Thermo Scientific™
5116-1300 |
0°C | 3 x 4 | 12 Tubes (12 to 13 mm dia.) |
464.00 €
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||
10253921
![]() |
Thermo Scientific™
5116-1600 |
0°C | 3 x 4 | 12 Tubes (16 to 17 mm dia.) |
465.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
10089140
![]() |
Thermo Scientific™
DS5116-0012 |
0°C | 3 x 4 | 12 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL) |
220.00 €
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||
10778391
![]() |
Thermo Scientific™
DS5116-0032 |
0°C | 4 x 8 | 32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL) |
425.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||
10741792
![]() |
Thermo Scientific™
DS5114-0012 |
-20°C | 3 x 4 | 12 Cryogenic Vials (0.2 to 1.5 mL) |
263.00 €
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||
Description
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
- Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
- Holds 12, 20 or 32 tubes
- Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
- Printed grid on one-way keyed lid for proper orientation and sample identification
- Molded of impact-resistant polycarbonate with locking handle for long-term use
- Stackable to save space
Specifications
Polycarbonate | |
Benchtop Cooler |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Benchtop Coolers