missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ Benchtop pH Meters

Compact unit that provides accurate and reliable readings.

Brand:  Jenway™ 35459-10

 View more versions of this product

Product Code. 12421240

  • 1100.00 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Supports one, two, or three point pH calibration
  • Up to three decimal place resolution for accurate readings
Specifications

Specifications

Advanced
±0.5°C/±1°F
-2 to 20
-1999.9 to 19.999
Advanced Digital pH Meter Kit w/GLP, glass electrode, ATC, buffers; 120 V
GLP, glass electrode, ATC, buffers
±0.003
0.001, 0.01, 0.1
LCD, 1/8 VGA Monochrome Backlit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers