Learn More
Lenz Laborglasinstrumente™ Bent Adapter
Allows for tubing connections. Lenz Laborglasinstrumente™ Bent Adapters feature a bent glass olive at 90°.
Brand:  Lenz Laborglasinstrumente™ 05095001
 View more versions of this product
Product Code. 11816084
  • 14.00 € / Each
Explore more special offers
Description
Description
  • Constructed from DURAN™ tubing
  • With NS socket
  • Bore: 2.5 mm
Specifications
Specifications
Bent Cone Adapter
Duran Glass
NS 14/23