Learn More
Corning™ Bent Discharge Tubes
Corning™ Bent Discharge Tubes for quick and precise dispensing of acids, bases and solvents
Brand:  Corning™ 6879
 View more versions of this product
Product Code. 16882120
  • 14.48 € / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bent
  • Includes protective sleeve
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bent
FEP, Polypropylene
Quick and precise dispensing of acids, bases and solvents
Protective Sleeve
Discharge Tube
2.5 to 10 mL
2.5 to 10 mL Bottle Top Dispenser
80 mm