Learn More
Benzeneboronic acid, 98+%, Thermo Scientific Chemicals
53.06 € - 664.31 €
Chemical Identifiers
CAS | 98-80-6 |
---|---|
Molecular Formula | C6H7BO2 |
Molecular Weight (g/mol) | 121.93 |
MDL Number | MFCD00002103 |
InChI Key | HXITXNWTGFUOAU-UHFFFAOYSA-N |
Synonym | benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 |
PubChem CID | 66827 |
ChEBI | CHEBI:44923 |
IUPAC Name | phenylboronic acid |
SMILES | OB(O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11422397
|
Thermo Scientific Alfa Aesar
A14257.09 |
10 g |
53.06 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432397
|
Thermo Scientific Alfa Aesar
A14257.18 |
50 g |
173.87 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442397
|
Thermo Scientific Alfa Aesar
A14257.30 |
250 g |
664.31 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Avoid moisture exposure.
Chemical Identifiers
98-80-6 | |
121.93 | |
HXITXNWTGFUOAU-UHFFFAOYSA-N | |
66827 | |
phenylboronic acid |
C6H7BO2 | |
MFCD00002103 | |
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
CHEBI:44923 | |
OB(O)C1=CC=CC=C1 |
Specifications
98-80-6 | |
C6H7BO2 | |
970972 | |
14,1068 | |
Slightly soluble in water. | |
OB(O)C1=CC=CC=C1 | |
121.93 | |
CHEBI:44923 | |
≥98% |
214°C to 219°C | |
MFCD00002103 | |
Hygroscopic | |
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
HXITXNWTGFUOAU-UHFFFAOYSA-N | |
phenylboronic acid | |
66827 | |
121.93 | |
Benzeneboronic acid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264-P270-P301+P312-P304+P312-P305+P351+P338-P312-P330-P501c
H302
missing translation for 'einecsNumber' : 202-701-9
missing translation for 'rtecsNumber' : CY8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Benzeneboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Benzeneboronic acid
RUO – Research Use Only