missing translation for 'onlineSavingsMsg'
Learn More
Benzofurazan-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals Product Code.: 11436660

Benzofurazan-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11436660
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11436660

Brand: Thermo Scientific Alfa Aesar H29358.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073355-14-2
Molecular Formula C12H15BN2O3
Molecular Weight (g/mol) 246.07
MDL Number MFCD09842717
InChI Key MQXANAGMQAYTMV-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo c 1,2,5 oxadiazole, benzofurazan-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzofurazan-5-boronic acid, pinacol ester, benzo c 1,2,5 oxadiazole-5-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzofurazan, benzo c 1,2,5 oxadiazole-5-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,1,3-benzoxadiazole, benzo c 1,2,5 oxadiazol-5-ylboronic acid pinacol ester, amtb001
PubChem CID 46738004
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzoxadiazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=NON=C2C=C1

Specifications

Melting Point 87°C to 88°C
Quantity 5 g
Formula Weight 246.07
Percent Purity 97%
Chemical Name or Material Benzofurazan-5-boronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10137498 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberH29358.03,H29358.06,H29358.MD
Lot Number10137498 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29358.03,H29358.06,H29358.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzofurazan-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.