missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 18 HPLC Columns, 3μm Particle Size
Available in limited dimensions. Please contact Customer Service for more details.
Brand: Thermo Scientific™ 71503-102130
911.68 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo ScientificTM BetaBasicTM 18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
LC/MS, APCI | |
3 μm | |
150 Å | |
L1 | |
Beta | |
13% | |
Yes |
2.1 mm | |
100 mm | |
2 to 12 | |
200 m²/g | |
C18 | |
Silica | |
Reversed Phase | |
Reversed Phase |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction