missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 18 HPLC Columns, 3μm Particle Size
Available in limited dimensions. Please contact Customer Service for more details.
727.26 € - 918.79 €
Specifications
Column Format | Analytical Column |
---|---|
For Use With | LC/MS, APCI |
Particle Size | 3 μm |
pH | 2 to 12 |
Pore Size | 150 Å |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
10487994
|
Thermo Scientific™
71503-034630 |
30 mm | 4.6 mm |
805.38 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
10210765
|
Thermo Scientific™
71503-052130 |
50 mm | 2.1 mm |
843.60 €
Each |
In Stock
Log in to see stock. |
||||
10456694
|
Thermo Scientific™
71503-054030 |
50 mm | 4 mm |
809.03 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
10270805
|
Thermo Scientific™
71503-054630 |
50 mm | 4.6 mm |
805.38 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
||||
10426684
|
Thermo Scientific™
71503-102130 |
100 mm | 2.1 mm |
916.24 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
||||
10477483
|
Thermo Scientific™
71503-104630 |
100 mm | 4.6 mm |
875.46 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||||
10360744
|
Thermo Scientific™
71503-152130 |
150 mm | 2.1 mm | N/A | |||||
10291235
|
Thermo Scientific™
71503-154630 |
150 mm | 4.6 mm |
918.79 €
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
Description
Thermo ScientificTM BetaBasicTM 18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
3 μm | |
150 Å | |
L1 | |
Silica | |
Reversed Phase | |
Reversed Phase |
LC/MS, APCI | |
2 to 12 | |
200 m²/g | |
C18 | |
13% | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title