Learn More
Thermo Scientific™ BetaSil™ Silica Guard Cartridges
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
Brand: Thermo Scientific™ 70005-013001
Description
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
Thermo Scientific™ BETASIL™ Silica HPLC columns are based on a highly pure, 100Å silica designed for small molecule analysis. BETASIL Silica columns are used for normal phase separations. These non-bonded silica columns are a powerful and efficient tool for separating non-polar and moderately polar compounds.
- 100Å high purity silica
- Used for the separation of non-polar and moderately polar compounds.
Specifications
2 to 9 | |
10 mm | |
3 mm | |
HPLC Column | |
L3 |
5 μm | |
100Å | |
3 mm | |
No | |
BetaSil |