Learn More
Thermo Scientific™ BetaSil™ Silica Guard Cartridges
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
Brand:  Thermo Scientific™ 70005-011001
 View more versions of this product
Product Code. 10514855
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
  • 100&Angst; silica
  • High surface area
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
1 mm I.D.
BetaSil
10 mm
1 mm