Learn More
Keystone Cleanroom Products BHD Autoclavable Wrapper
BIOPHARMAGREENER_CHOICE
When secured with autoclave tape or elasticized bands, these BHD Autoclavable Wrappers provide an adequate microbial barrier from external sources of contamination.
836.00 €
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance
Closure Type None
Color Blue
Disposable Single-use
View More Specs
Products 5
Product Code Brand Width (English) Length (English) Price Quantity & Availability  
Product Code Brand Width (English) Length (English) Price Quantity & Availability  
17798177
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BHDSHEET01.5X01.5
1.5 in. 1.5 in.
836.00 €
Pack of 3000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
17708187
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BHDSHEET0202
2 in. 2 in.
836.00 €
Pack of 3000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
17718187
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BHDSHEET0303
3 in. 3 in.
836.00 €
Pack of 3000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
17728187
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BHDSHEET0404
4 in. 4 in.
836.00 €
Pack of 3000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
17738187
BIOPHARMAGREENER_CHOICE
View Documents
Keystone Cleanroom Products
BHDSHEET0606
6 in. 6 in.
836.00 €
Pack of 3000
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
Description
Description
  • Designed for steam sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO Certified Cleanrooms
  • Certificate of Conformance included with shipment
Specifications
Specifications
Autoclavable
None
Single-use
Cellulose
Yes
Certificate of Conformance
Blue
Gamma, EtO, or Steam Sterilization
Non-sterile
Up to 135°C
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Keystone Cleanroom Products BHD Autoclavable Wrapper