missing translation for 'onlineSavingsMsg'
Learn More
Honeywell™ Bilson 303L Foam Earplug Product Code.: 12676113
Add to basket

Honeywell™ Bilson 303L Foam Earplug

Product Code. 12676113
Yellow/White
Click to view available options
Yellow/White
This item is not returnable. View return policy

Product Code. 12676113

Brand: Honeywell™ 1007192

Edge
Add to basket
Add to basket
This item is not returnable. View return policy

Soft foam earplugs provide superior long-wearing comfort and solid performance.

  • Soft foam for comfort and performance
  • Smooth, soil-resistant surface prevents dirt or other substances from accumulating on skin or entering ear canal
  • Ideal for workers who need to block out loud noise and demand the best protection and comfort available in an earplug
  • Europe's most used polyurethane earplug
  • Superior noise-blocking performance in a single-use earplug
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow, White
Certifications/Compliance (Europe) EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: 941057, EC Attestation: 9404815
Noise Reduction Rating (NRR) SNR: 33 dB
Plug Shape Bullet
Product Type Single-use earplug
Disposable Single-use
Material Polyurethane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell™ Bilson 303L Foam Earplug >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.