Learn More
Hirschmann™ BIO-SILICO™ Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with coarse bubbles for preparation, filling and sterilization of culture media.
Brand:  Hirschmann™ 8905922
 View more versions of this product
Product Code. 10007792
  • 82.10 € / Pack of 10
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Coarse pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications
Specifications
BIO-SILICO, N22
For The Preparation And Sterilization Of Culture Media
18 mm
Orange
24 mm
Silicone Stopper
18-21 mm
30 mm
10/Pack
18 mm
Silicone