missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Biobased Flat Tray

Biobased Flat Tray has dimensions suitable for refrigerators.

Brand:  Gilac™ G152321

 View more versions of this product

Product Code. 17918951

  • 15.65 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flat tray in biobased material.
  • Dimensions suitable for refrigerators.
  • Flat tray can be used as:
    • defrosting trays thanks to its dividing grid that separates the product from the defrosting water.
    • preparation container for products that drip.
    • ingredients containers.
    • containers for preserving comparison samples.
    • Fermenting and decanting containers.
  • Nesting and stacking with lid. Traceability management thanks to a contents identification label (IML).
  • 3 volumes available: 3 L, 5 L, 8 L.
  • Biobased flat trays are white and dye-free.
  • Biobased material is made from natural renewable resources instead of fossil resources.
  • As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety.
Specifications

Specifications

5 L
75 mm
HDPE
285 mm
3 in.
0.8 gal.
White
437 mm
Laboratory Tray
13.7 in.
11.2 in.
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers