missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Biobased Flat Tray

Biobased Flat Tray has dimensions suitable for refrigerators.

Brand:  Gilac™ G152221

 View more versions of this product

Product Code. 17938551

  • 11.10 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Flat tray in biobased material.
  • Dimensions suitable for refrigerators.
  • Flat tray can be used as:
    • defrosting trays thanks to its dividing grid that separates the product from the defrosting water.
    • preparation container for products that drip.
    • ingredients containers.
    • containers for preserving comparison samples.
    • Fermenting and decanting containers.
  • Nesting and stacking with lid. Traceability management thanks to a contents identification label (IML).
  • 3 volumes available: 3 L, 5 L, 8 L.
  • Biobased flat trays are white and dye-free.
  • Biobased material is made from natural renewable resources instead of fossil resources.
  • As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety.
Specifications

Specifications

3 L
70 mm
HDPE
230 mm
2.8 in.
0.8 gal.
White
347 mm
Laboratory Tray
13.7 in.
9.1 in.
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers