Learn More
Nitritex™ BioClean-D Disposable Laboratory Coats
Wear Nitritex™ BioClean-D Disposable Laboratory Coats to protect against laboratory contamination.
260.00 € - 263.00 €
Specifications
Product Type Controlled Environments Lab Coat
Material Double Laminate Microporous Fabric
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12912255
View Documents
Nitritex™
BDLC-S
Small
260.00 €
Pack of 30
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
12942255
View Documents
Nitritex™
BDLC-XL
X-Large
263.00 €
Pack of 30
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
12922255
View Documents
Nitritex™
BDLC-M
Medium
263.00 €
Pack of 30
In Stock
to see stock.
Edge
Add to basket
12932255
View Documents
Nitritex™
BDLC-L
Large
260.00 €
Pack of 30
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
  • Open cuff style
  • Three pockets
  • CleanTough™ Fabric
  • Disposable, packaged in bag
  • Spunbond Polypropylene
Specifications
Specifications
Controlled Environments Lab Coat
Open
Non-sterile
ISO 9001:2015, ASTM F739-12
One piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 30 outer bags per lined carton (30 pieces)
Double Laminate Microporous Fabric
Single-use
3
(EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Spunbond Polypropylene