missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Disposal Bags with/without Warning Label

Economical, safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.

Brand:  Bel-Art™ F13162-0009

 View more versions of this product

Product Code. 11372154

  • 179.05 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Economical, safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes

  • These transparent, polypropylene plastic biohazard bags are 0.03mm (1.5 mil) thick
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners

Specifications

Specifications

Biohazard Bag
ASTM D1709-98
Single-use
Biohazard Symbol with Sterilization Patch
1.5 mm
36 in.
24 in.
Autoclavable
Clear
Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Polypropylene
36 x 24 in. (91 x 61 cm)
91 cm
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers