missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Biopur™ epT.I.P.S.™ Singles
Eppendorf™ Biopur™ epT.I.P.S.™ Singles are the perfect combination that results in reduced-force tip attachment and ejection with complete sealing.
34.80 € - 36.29 €
Specifications
Filtered | Non-filtered |
---|---|
Sterility | Sterile |
Color | Natural |
Autoclavable | Autoclavable |
Model | epT.I.P.S.™ Singles |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
15851475
|
Eppendorf™
0030010027 |
0.1 to 20 μL | 100 tips, individually packed |
34.80 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15861475
|
Eppendorf™
0030010043 |
2 to 200 μL | 100 tips, individually packed |
36.29 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
15871475
|
Eppendorf™
0030010060 |
50 to 1000 μL | 100 tips, individually packed |
36.29 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Ensured PCR clean, sterile, pyrogen-, ATP- and DNA-free
- For use in pharmaceutical and food industry, molecular biology and cell technology
- Batch number and expiration date on each blister package
- Each production lot inspected continuously by an independent laboratory
Specifications
Non-filtered | |
Natural | |
epT.I.P.S.™ Singles | |
Singles | |
epT.I.P.S. |
Sterile | |
Autoclavable | |
Biopur™ | |
100 tips, individually packed |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title