missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals
34.80 € - 112.00 €
Chemical Identifiers
CAS | 135-57-9 |
---|---|
Molecular Formula | C26H20N2O2S2 |
Molecular Weight (g/mol) | 456.578 |
MDL Number | MFCD00043806 |
InChI Key | ZHMIOPLMFZVSHY-UHFFFAOYSA-N |
Synonym | bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide |
PubChem CID | 67271 |
IUPAC Name | N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide |
SMILES | C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398615
|
Thermo Scientific Alfa Aesar
L12110.14 |
25 g |
34.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308625
|
Thermo Scientific Alfa Aesar
L12110.22 |
100 g |
112.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
135-57-9 | |
456.578 | |
ZHMIOPLMFZVSHY-UHFFFAOYSA-N | |
67271 | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4 |
C26H20N2O2S2 | |
MFCD00043806 | |
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide | |
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide |
Specifications
135-57-9 | |
C26H20N2O2S2 | |
721443 | |
ZHMIOPLMFZVSHY-UHFFFAOYSA-N | |
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide | |
67271 | |
97% |
140°C to 143°C | |
MFCD00043806 | |
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4 | |
456.578 | |
456.59 | |
Bis(2-benzamidophenyl) disulfide |
Safety and Handling
missing translation for 'einecsNumber' : 205-201-9
missing translation for 'rtecsNumber' : CV8700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only