missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)titanium dichloride, 99+%, Thermo Scientific Chemicals

27.94 € - 228.00 €

Chemical Identifiers

CAS 1271-19-8
Molecular Formula C10H10Cl2Ti-2
Molecular Weight (g/mol) 248.957
MDL Number MFCD00003723
InChI Key MKNXBRLZBFVUPV-UHFFFAOYSA-L
Synonym Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
PubChem CID 124040768
IUPAC Name cyclopenta-1,3-diene;titanium(2+);dichloride
SMILES [CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040386.04
2 g
27.94 €
2g
Please to purchase this item. Need a web account? Register with us today!
11353958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040386.09
10 g
55.28 €
10g
Please to purchase this item. Need a web account? Register with us today!
11383958
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040386.18
50 g
228.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This metallocene is a common reagent in organometallic and organic synthesis. Titanocene dichloride is used to prepare titanocene pentasulfide. Used as an anticancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

1271-19-8
248.957
MKNXBRLZBFVUPV-UHFFFAOYSA-L
124040768
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
C10H10Cl2Ti-2
MFCD00003723
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
cyclopenta-1,3-diene;titanium(2+);dichloride

Specifications

1271-19-8
Red
Odorless
MFCD00003723
14,9482
Reacts in moist air and in water to form HCl . Slightly hydrolysis in water.
[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Cl-].[Ti+2]
248.957
249
Crystalline
∼285°C (decomposition)
1.60 g/mL
C10H10Cl2Ti-2
Air and moisture sensitive
Dichlorobis(cyclopentadienyl)titanium; Titanocene dichloride
MKNXBRLZBFVUPV-UHFFFAOYSA-L
cyclopenta-1,3-diene;titanium(2+);dichloride
124040768
≥99%
Bis(cyclopentadienyl)titanium dichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302-H332-H315-H319-H335
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 215-035-9

missing translation for 'rtecsNumber' : XR2050000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only