missing translation for 'onlineSavingsMsg'
Learn More
Bis(ethylenedithio)tetrathiafulvalene, Thermo Scientific Chemicals Product Code.: 11378368

Bis(ethylenedithio)tetrathiafulvalene, Thermo Scientific Chemicals

Product Code. 11378368
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11378368

Brand: Thermo Scientific Alfa Aesar 039557.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a component for organic superconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66946-48-3
Molecular Formula C10H8S8
Molecular Weight (g/mol) 384.65
MDL Number MFCD00059710
InChI Key LZJCVNLYDXCIBG-UHFFFAOYSA-N
Synonym bis ethylenedithio tetrathiafulvalene, bedt-ttf, 2-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5h,6h-1,3 dithiolo 4,5-b 1,4 dithiine, bis ethylenedithio tetrathiofulvalene, bis ethylenedithiolo tetrathiafulvalene, bis-ethylenedithio tetrathiafulvalene, bis ethylenedithiolo tetrathiafulvalene 250mg, bi 5,6-dihydro-1,3-dithiolo 4,5-b 1,4-dithiine-2-ylidene, 2-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiin-2-ylidene-5,6-dihydro-1,3 dithiolo 4,5-b 1,4 dithiineShow MoreShow Less
PubChem CID 633432
IUPAC Name 2-(5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene)-5,6-dihydro-[1,3]dithiolo[4,5-b][1,4]dithiine
SMILES C1CSC2=C(SC(S2)=C2SC3=C(S2)SCCS3)S1

Specifications

Melting Point 238°C (decomposition)
Quantity 1 g
Solubility Information Soluble in organic solvents.
Formula Weight 384.68
Physical Form Powder
Chemical Name or Material Bis(ethylenedithiolo)tetrathiafulvalene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ24K050 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number039557.03,039557.MD
Lot NumberU08E018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number039557.03,039557.MD
Lot NumberD25X028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039557.03,039557.MD
Lot NumberX27C018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039557.03,039557.MD
Lot NumberH13U002 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039557.03,039557.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bis(ethylenedithio)tetrathiafulvalene, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.