missing translation for 'onlineSavingsMsg'
Learn More

Bis(heptafluoroisopropyl)ketone, 97%, Thermo Scientific Chemicals

62.50 € - 231.00 €

Chemical Identifiers

CAS 813-44-5
Molecular Formula C7F14O
Molecular Weight (g/mol) 366.054
MDL Number MFCD00042087
InChI Key GRVMOMUDALILLH-UHFFFAOYSA-N
Synonym bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat
PubChem CID 69941
IUPAC Name 1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
SMILES C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339345
View Documents
Thermo Scientific Alfa Aesar
L16708.03
1 g
62.50 €
1g
Please to purchase this item. Need a web account? Register with us today!
11349345
View Documents
Thermo Scientific Alfa Aesar
L16708.06
5 g
231.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

813-44-5
366.054
GRVMOMUDALILLH-UHFFFAOYSA-N
69941
C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F
C7F14O
MFCD00042087
bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat
1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one

Specifications

813-44-5
72°C to 73°C
MFCD00042087
1916532
GRVMOMUDALILLH-UHFFFAOYSA-N
1,1,1,2,4,5,5,5-octafluoro-2,4-bis(trifluoromethyl)pentan-3-one
69941
97%
1.661
C7F14O
UN2810
bis perfluoroisopropyl ketone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl pentan-3-one, bis perfluorisopropyl ketone, bis heptafluoroisopropyl ketone, 3-pentanone, 1,1,1,2,4,5,5,5-octafluoro-2,4-bis trifluoromethyl, perfluoroisopropyl ketone, bis heptafluoroisopropyl methanone, cf3 2cfc o cf cf3 2, 3-hexanone, 1,1,1,2,4,4,5,5,6,6,6-undecafluoro-2-trifluoromethyl, tert-butyl 2-oxo-1-pyrrolidinecarboxylat
C(=O)(C(C(F)(F)F)(C(F)(F)F)F)C(C(F)(F)F)(C(F)(F)F)F
366.054
366.06
Bis(heptafluoroisopropyl)ketone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 212-384-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only