Learn More
Melford bis-MSB Scintillation Grade
High solubility secondary fluor.
Brand:  Melford 111077
 View more versions of this product
Product Code. 30017920
  • 336.00 € / 25g
Explore more special offers
Description
Description
High solubility secondary fluor.
High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A
Specifications
Specifications
bis-MSB, Scintillation Grade
25 g