missing translation for 'onlineSavingsMsg'
Learn More

Melford bis-MSB Scintillation Grade

High solubility secondary fluor.

113.00 € - 336.00 €

Specifications

Chemical Name or Material bis-MSB, Scintillation Grade
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30017920
View Documents
Melford
111077
25 g
336.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
30017919
View Documents
Melford
111076
5 g
113.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

High solubility secondary fluor.

High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A
Specifications

Specifications

bis-MSB, Scintillation Grade
Videos
SDS
Documents

Documents

Product Certifications