Learn More
Bis(tri-n-butyltin) oxide, 97%, Thermo Scientific Chemicals
45.23 € - 456.00 €
Chemical Identifiers
CAS | 56-35-9 |
---|---|
Molecular Formula | C24H58OSn2 |
Molecular Weight (g/mol) | 600.15 |
MDL Number | MFCD00009418 |
InChI Key | NZQFYJHTWVEUPP-UHFFFAOYSA-N |
Synonym | tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biomet |
PubChem CID | 16682746 |
ChEBI | CHEBI:81543 |
IUPAC Name | tributyl(tributylstannyloxy)stannane |
SMILES | O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11480773
|
Thermo Scientific Alfa Aesar
A13242.22 |
100 g |
45.23 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11490773
|
Thermo Scientific Alfa Aesar
A13242.36 |
500 g |
150.75 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400783
|
Thermo Scientific Alfa Aesar
A13242.0E |
2500 g | N/A | ||||||
Description
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Chemical Identifiers
56-35-9 | |
600.15 | |
NZQFYJHTWVEUPP-UHFFFAOYSA-N | |
16682746 | |
tributyl(tributylstannyloxy)stannane |
C24H58OSn2 | |
MFCD00009418 | |
tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biomet | |
CHEBI:81543 | |
O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC |
Specifications
56-35-9 | |
1.172 | |
168°C (334°F) | |
C24H58OSn2 | |
1.4865 | |
UN2788 | |
tributyltin oxide, tbto, bis tributyltin oxide, hexabutyldistannoxane, distannoxane, hexabutyl, lastanox q, bis tri-n-butyltin oxide, butinox, biomet | |
NZQFYJHTWVEUPP-UHFFFAOYSA-N | |
tributyl(tributylstannyloxy)stannane | |
16682746 | |
596.07 | |
Bis(tri-n-butyltin) oxide |
-45°C | |
179°C to 180°C (2 mmHg) | |
Irritating | |
[(CH3CH2CH2CH2)3Sn]2O | |
MFCD00009418 | |
745057 | |
Soluble in water (0.071g/L) | |
O.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC | |
600.15 | |
CHEBI:81543 | |
97% |
Safety and Handling
GHS H Statement
H301-H311-H360-H372-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H335-H360FD-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 200-268-0
missing translation for 'rtecsNumber' : JN8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.