missing translation for 'onlineSavingsMsg'
Learn More
Bismuth needles, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11354228

Bismuth needles, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11354228
2 kg
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11354228

Brand: Thermo Scientific Alfa Aesar 041485.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: Approx. 1.5-3.0mm diameter; Various lengths

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]

Specifications

Color Gray
Appearance Gray needles
Density 9.79g/cm3
Assay Percent Range 99.99% (Metals basis)
Quantity 2 kg
Odor Odorless
Form Needle
Chemical Name or Material Bismuth

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR09I027 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number041485.36,041485.A3,041485.22,041497.30
Lot NumberY08H078 Certificate TypeCertificate of Analysis Date15/06/2022 Catalog Number041485.36,041485.A3,041485.22,041497.30
Lot NumberZ13E042 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number041485.36,041485.A3,041485.22,041497.30
Lot NumberF11W005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041485.36,041485.A3,041485.22,041497.30
Lot NumberX20H036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041485.36,041485.A3,041485.22,041497.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bismuth needles, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.