missing translation for 'onlineSavingsMsg'
Learn More

Bismuth powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

60.00 € - 549.74 €

Chemical Identifiers

CAS 7440-69-9
Molecular Formula Bi
Molecular Weight (g/mol) 208.98
MDL Number MFCD00134033
InChI Key JCXGWMGPZLAOME-UHFFFAOYSA-N
Synonym bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
PubChem CID 5359367
ChEBI CHEBI:33301
IUPAC Name bismuth
SMILES [Bi]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11341158
SDS View Documents
Thermo Scientific Alfa Aesar
010110.22
100 g
60.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
11351158
SDS View Documents
Thermo Scientific Alfa Aesar
010110.36
500 g
190.95 €
500g
Please to purchase this item. Need a web account? Register with us today!
11361158
SDS View Documents
Thermo Scientific Alfa Aesar
010110.A3
2 kg
549.74 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]

Specifications

7440-69-9
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
[Bi]
208.98
CHEBI:33301
−100 Mesh
Apparent density: 3.8g/cm3 to 4.6g/cm3
(metals basis)
Bi
UN3089
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
5359367
99.5%
Odorless
Powder
Bismuth powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-177-4

missing translation for 'rtecsNumber' : EB2600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Bismuth powder

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use to extinguish.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.