Learn More
Thermo Scientific™ Black 96-Well Immuno Plates

Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with these black, 96-well immuno plates, available in a choice of three surface types.
Brand: Thermo Scientific™ 437111
275.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Perform quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ Black 96-Well Immuno Plates. Featuring a choice of three surface types and optimal binding, these plates minimize background in fluorescence reading.
Features:
- Plate has SBS footprint — fits standard equipment
- Flat-bottom wells
- Certified binding homogeneity/reproducibility
Choice of surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) are for absorption of hydrophobic molecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
- Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules.
Compatible with:
Compatible with lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country

Specifications
Hydrophilic | |
Without Lid | |
8 x 12 | |
Adsorption | |
Microplate | |
96 | |
MaxiSorp | |
Fluorescence | |
10 | |
Antibodies (Glycoproteins) |
Standard | |
Fluorescence | |
80 Cases | |
Black | |
Polystyrene | |
350 μL | |
Flat Bottom (F-well) | |
80 | |
None | |
F96 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
185187 | Certificate of Compliance | 28/02/2025 | 437111 |
184515 | Certificate of Compliance | 11/12/2024 | 437111 |
185048 | Certificate of Compliance | 10/12/2024 | 437111 |
181102 | Certificate of Compliance | 03/12/2024 | 437111 |
164664 | Certificate of Compliance | 03/12/2024 | 437111 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.