missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Counting Chambers, Neubauer Pattern, BLAUBRAND™, double ruling
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
Brand: BRAND™ 718605
59.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
- The ruling shows 9 large squares of 1 mm2 each.
- The four large squares in the corners marked "L" for counting leucocytes are each subdivided into 16 squares with 0.25 mm sides.
- The large square in the center is subdivided into 16 squares with 0.25 mm sides.
- Each group square consists of 16 mini squares with 0.05 mm sides, each having an area of 0.0025 mm2.
- The 5 group squares in the large central square are used for counting thrombocytes and erythrocytes.
- Contrary to the more advanced Improved Neubauer counting chambers, the counting area of each group square is limited by the outer one of the triple boundary lines.

Specifications
Counting Device | |
0.1mm | |
Without spring clips |
1Each | |
Counting leucocytes, thrombocytes and erythrocytes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ Counting Chambers, Neubauer Pattern, BLAUBRAND™, double ruling > Without Spring Clips
Spot an opportunity for improvement?Share a Content Correction