missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blocker™ BLOTTO in TBS

Block membranes, plates and other samples with this ready-to-use 5% solution of nonfat powdered milk in TBS for Western blot, ELISA and other detection methods.

Brand:  Thermo Scientific™ 37530

165.01 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10016913

  • 184.00 € / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Blocker BLOTTO Blocking Buffer is a ready-to-use 5% solution of nonfat powdered milk in Tris-buffered saline for using in Western blotting, ELISA, immunohistochemistry and other detection methods.

Highlights:

  • Popular – nonfat milk has been used for many years in a variety of protein methods, although it is not recommended for avidin-based techniques because it contains some endogenous biotin

  • Convenient – supplied as a ready-to-use 1X TBS solution; can be diluted as needed

  • Easy to use – formulated with anti-foaming agent and thimerosal-free preservative

  • Flexible – may be used for multiple applications, including as a diluent for antibody

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blocker BLOTTO
Liquid
Blocker™
TBS
Store at 4°C.
1 L
Western Blot, ELISA, Immunohistochemistry (IHC)
1X
5% non-fat milk
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK404391 Certificate TypeCertificate of Analysis Date24/12/2024 Catalog Number37530
Lot NumberZJ403683 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number37530
Lot NumberZJ403419 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number37530
Lot NumberZH401604 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number37530
Lot NumberZI402176 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number37530
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Blocker™ BLOTTO in TBS > 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.