Learn More
Keystone Cleanroom Products Blue SMS Sterilization Wrap, 50 gsm
BIOPHARMAGREENER_CHOICE
SMS sterilization wraps provide a microbial barrier and are ideal for pharmaceutical production applications. They are designed for use in steam, ethylene oxide, and hydrogen peroxide sterilization processes.
Brand:  Keystone Cleanroom Products KCRP-300-0909
 View more versions of this product
Product Code. 17811481
  • 290.00 € / Pack of 1000
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured and packaged in a Certified ISO 8 cleanroom
  • Clean - no cellulose
  • Strong, rip and tear resistant 50 gsm material
  • Soft and drape-able fabric
  • An extensive range of sizes available means less cutting
  • Hydrophobic barrier allows sterilization gasses but prevents liquid and microbial penetration
  • Sub packed into sealed labeled poly bags with Lot Number and Date of Manufacture for complete traceability
  • Certificate of Conformance sent with each shipment
  • Made in the USA
Specifications
Specifications
Sterilization Wrap (Heavy-duty)
COC
SMS Fabric
Yes
22.9 x 22.9 cm (9 x 9 in.)
Autoclavable
Blue
Non-sterile
121°C to 125°C